missing translation for 'onlineSavingsMsg'
Learn More

SC 26196, Tocris Bioscience™

Selective Δ6 desaturase inhibitor

Marke:  Tocris (Bio-Techne) 418910

Weitere Details : Netto-Gewicht : 0.01000kg

Menge. 10mg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 13414314

  • 224.41 € / 10 Milligramm


Voraussichtlicher Liefertermin 04-06-2019
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Selective fatty acid desaturase 2 (FADS2, also known as Δ 6 desaturase) inhibitor (IC 50 = 0.2 μM in vitro). Displays selectivity over FADS1 and stearoly-CoA 9-desaturase (Δ 5 desaturase and SCD-1/Δ 9 desaturase, respectively) (IC 50 values are >200 μM in vitro). Exhibits anti-inflammatory properties in a mouse edema model. Eliminates CSCs from ovarian cancer cell lines and inhibits sphere formation in vitro.
Spezifikation

Chemische Identifikatoren

218136-59-5
423.564
α, α-Diphenyl-4-[(3-pyridinylmethylene)amino]-1-piperazinepentanenitrile
2,2-diphenyl-5-[4-(pyridin-3-ylmethylideneamino)piperazin-1-yl]pentanenitrile
C27H29N5
QFYKXKMYVYOUNJ-UHFFFAOYSA-N
71752132
C1CN(CCN1CCCC(C#N)(C2=CC=CC=C2)C3=CC=CC=C3)N=CC4=CN=CC=C4

Spezifikation

SC 26196
218136-59-5
α, α-Diphenyl-4-[(3-pyridinylmethylene)amino]-1-piperazinepentanenitrile
C1CN(CCN1CCCC(C#N)(C2=CC=CC=C2)C3=CC=CC=C3)N=CC4=CN=CC=C4
423.564
423.55
10mg
C27H29N5
QFYKXKMYVYOUNJ-UHFFFAOYSA-N
2,2-diphenyl-5-[4-(pyridin-3-ylmethylideneamino)piperazin-1-yl]pentanenitrile
71752132
>98%
Videos