missing translation for 'onlineSavingsMsg'
Learn More

SC 57461A, Tocris Bioscience™

Selective inhibitor of LTA4 hydrolase

Marke:  Tocris (Bio-Techne) 310710

Weitere Details : Netto-Gewicht : 0.01000kg

Menge. 10mg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 13412454

  • 163.68 € / 10 Milligramm


Voraussichtlicher Liefertermin 30-04-2019
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Potent and selective inhibitor of LTA 4 hydrolase. Does not inhibit other enzymes of the arachidonic acid cascade including COX-1, COX-2, LTC 4 synthase and 5-lipoxygenase. Potently inhibits LTB 4 production in whole blood (IC 50 = 49 nM). Orally active.
Spezifikation

Chemische Identifikatoren

423169-68-0
363.882
sc-57461 hydrochloride, n-methyl-n-3-4-phenylmethyl phenoxy propyl-, a-alanine hydrochloride, 3-methyl 3-4-phenylmethyl phenoxy propyl amino propanoic acid, 3-3-4-benzylphenoxy propyl methyl amino propanoic acid hydrochloride, 3-3-4-benzylphenoxy propyl methyl amino propanoic acid hydrochloride, n-methyl-n-3-4-phenylmethyl phenoxy propyl-beta-alanine, sc-57461a hplc, n-methyl-n-3-4-phenylmethyl phenoxy propyl, a-alanine hydrochloride, n-methyl-n-3-4-phenylmethyl phenoxy propyl beta-alanine hydrochloride, n-methyl-n-3-4-phenylmethyl phenoxy propyl-?-alanine hydrochloride
3-[3-(4-benzylphenoxy)propyl-methylamino]propanoic acid;hydrochloride
C20H26ClNO3
SBBJJLDNKNNWFJ-UHFFFAOYSA-N
9820433
CN(CCCOC1=CC=C(C=C1)CC2=CC=CC=C2)CCC(=O)O.Cl

Spezifikation

SC 57461A
423169-68-0
sc-57461 hydrochloride, n-methyl-n-3-4-phenylmethyl phenoxy propyl-, a-alanine hydrochloride, 3-methyl 3-4-phenylmethyl phenoxy propyl amino propanoic acid, 3-3-4-benzylphenoxy propyl methyl amino propanoic acid hydrochloride, 3-3-4-benzylphenoxy propyl methyl amino propanoic acid hydrochloride, n-methyl-n-3-4-phenylmethyl phenoxy propyl-beta-alanine, sc-57461a hplc, n-methyl-n-3-4-phenylmethyl phenoxy propyl, a-alanine hydrochloride, n-methyl-n-3-4-phenylmethyl phenoxy propyl beta-alanine hydrochloride, n-methyl-n-3-4-phenylmethyl phenoxy propyl-?-alanine hydrochloride
CN(CCCOC1=CC=C(C=C1)CC2=CC=CC=C2)CCC(=O)O.Cl
363.882
363.88
10mg
C20H26ClNO3
SBBJJLDNKNNWFJ-UHFFFAOYSA-N
3-[3-(4-benzylphenoxy)propyl-methylamino]propanoic acid;hydrochloride
9820433
>98%
Videos