missing translation for 'onlineSavingsMsg'
Learn More

SCH 23390 hydrochloride, Tocris Bioscience™

Standard selective D1-like antagonist; also 5-HT2C and 5-HT1C agonist and Kir3 channel blocker

Marke:  Tocris (Bio-Techne) 092550

Weitere Details : Netto-Gewicht : 0.01000kg

Menge. 50mg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 13418163

  • 691.16 € / 50 Milligramm


Voraussichtlicher Liefertermin 25-02-2019
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Potent dopamine receptor antagonist (Ki values are 0.2 nM and 0.3 nM at D1 and D5 receptor sub-types, respectively). Also an agonist at 5-HT1C and 5-HT2C receptors in vitro (Ki values are 6.3 nM and 9.3 nM respectively). Blocks quinpirole-induced Kir3 (GIRK) currents (EC50 = 268 nM) independently of receptors.
Spezifikation

Chemische Identifikatoren

125941-87-9
324.245
r +-sch-23390 hydrochloride, sch 23390 hydrochloride, sch-23390 hydrochloride, unii-4s8t3e2f4z, r-+-sch 23390 hydrochloride, sch 23390 hydrochloride, 5r-8-chloro-2,3,4,5-tetrahydro-3-methyl-5-phenyl-1h-3-benzazepin-7-ol hydrochloride, r +-7-chloro-8-hydroxy-3-methyl-1-phenyl-2,3,4,5-tetrahydro-1h-3-benzazepine hydrochloride, r-+-7-chloro-8-hydroxy-3-methyl-1-phenyl-2,3,4,5-tetrahydro-1h-3-benzazepine hydrochloride, sch23390 hydrochloride
(5R)-8-chloro-3-methyl-5-phenyl-1,2,4,5-tetrahydro-3-benzazepin-7-ol;hydrochloride
C17H19Cl2NO
OYCAEWMSOPMASE-XFULWGLBSA-N
11957535
CN1CCC2=CC(=C(C=C2C(C1)C3=CC=CC=C3)O)Cl.Cl

Spezifikation

SCH 23390 hydrochloride
125941-87-9
r +-sch-23390 hydrochloride, sch 23390 hydrochloride, sch-23390 hydrochloride, unii-4s8t3e2f4z, r-+-sch 23390 hydrochloride, sch 23390 hydrochloride, 5r-8-chloro-2,3,4,5-tetrahydro-3-methyl-5-phenyl-1h-3-benzazepin-7-ol hydrochloride, r +-7-chloro-8-hydroxy-3-methyl-1-phenyl-2,3,4,5-tetrahydro-1h-3-benzazepine hydrochloride, r-+-7-chloro-8-hydroxy-3-methyl-1-phenyl-2,3,4,5-tetrahydro-1h-3-benzazepine hydrochloride, sch23390 hydrochloride
CN1CCC2=CC(=C(C=C2C(C1)C3=CC=CC=C3)O)Cl.Cl
324.245
324.24
50mg
C17H19Cl2NO
OYCAEWMSOPMASE-XFULWGLBSA-N
(5R)-8-chloro-3-methyl-5-phenyl-1,2,4,5-tetrahydro-3-benzazepin-7-ol;hydrochloride
11957535
>98%
Videos