missing translation for 'onlineSavingsMsg'
Learn More

SCPx Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marke:  Proteintech 14397-1-AP-150UL

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 16883014

  • 329.85 € / 150 Mikroliter

Voraussichtliches Versanddatum: 25-06-2021

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

14397-1-AP antibody recognizes the full-length protein and the thiolase domain ofSCPx.

This gene encodes two proteins: sterol carrier protein X (SCPx) and sterol carrier protein 2 (SCP2), as a result of transcription initiation from 2 independently regulated promoters. The transcript initiated from the proximal promoter encodes the longer SCPx protein, and the transcript initiated from the distal promoter encodes the shorter SCP2 protein, with the 2 proteins sharing a common C-terminus. Evidence suggests that the SCPx protein is a peroxisome-associated thiolase that is involved in the oxidation of branched chain fatty acids, while the SCP2 protein is thought to be an intracellular lipid transfer protein. This gene is highly expressed in organs involved in lipid metabolism, and may play a role in Zellweger syndrome, in which cells are deficient in peroxisomes and have impaired bile acid synthesis. Alternative splicing of this gene produces multiple transcript variants, some encoding different isoforms. The full-length nature of all transcript variants has not been determined.
Spezifikation

Spezifikation

SCPx
0.27 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P11915, P22307, P32020
NLTP, NSL TP, SCP 2, SCP chi, SCP X, SCP2, SCPX, sterol carrier protein 2, Sterol carrier protein X
Rabbit
IgG
150 μL
-20°C
Primary
20280, 25541, 6342
Immunocytochemistry, Immunofluorescence, Western Blot
Unconjugated
SCP2
Liquid
SCP2
SCPx Fusion Protein Ag5895
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Zertifikate