missing translation for 'onlineSavingsMsg'
Learn More

SCRIB Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marke:  Proteintech 27083-1-AP-20UL

Weitere Details : Netto-Gewicht : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16885375

  • 112.71 € / 20 Mikroliter

Verfügbar ab
17-06-2021

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Description

Description

SCRIB is a cytoplasmic multimodular scaffold protein targeted to epithelial adherens junctions and neuronal presynaptic compartments. SCRIB and its orthologs in vertebrates and invertebrates participate in cell polarization.
Spécification

Spécification

SCRIB
0.5 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q14160, Q80U72
CRIB1, hScrib, KIAA0147, LAP4, Protein LAP4, Protein scribble homolog, SCRB1, SCRIB, SCRIB1, Scribble, scribbled homolog (Drosophila), Vartul
Rabbit
IgG
20 μL
-20°C
Primary
105782, 23513
Western Blot
Unconjugated
SCRIB
Liquid
Scrib
SCRIB Fusion Protein Ag25795
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Vidéos
FDS
Documents