missing translation for 'onlineSavingsMsg'
Learn More

Secretogranin V Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marke:  Proteintech 10761-1-AP-150UL

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 16887353

  • 329.85 € / 150 Mikroliter

Verfügbar ab
17-06-2021

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

Acts as a molecular chaperone for PCSK2/PC2, preventing its premature activation in the regulated secretory pathway. Binds to inactive PCSK2 in the endoplasmic reticulum and facilitates its transport from there to later compartments of the secretory pathway where it is proteolytically matured and activated. Also required for cleavage of PCSK2 but does not appear to be involved in its folding. Plays a role in regulating pituitary hormone secretion. The C-terminal peptide inhibits PCSK2 in vitro.
Spezifikation

Spezifikation

Secretogranin V
0.15 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P05408, P12961, P27682
7B2, Neuroendocrine protein 7B2, P7B2, Pituitary polypeptide, SCG5, Secretogranin 5, Secretogranin V, secretogranin V (7B2 protein), SGNE1, SgV
Rabbit
IgG
150 μL
-20°C
Primary
20394, 25719, 6447
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
SCG5
Liquid
SCG5
Secretogranin V Fusion Protein Ag1107
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation