missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
SEL1L3 Polyclonal specifically detects SEL1L3 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
Specifications
| Antigen | SEL1L3 |
| Anwendungen | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Klassifikation | Polyclonal |
| Konjugat | Unconjugated |
| Verdünnung | Immunocytochemistry/Immunofluorescence 1-4 μg/mL, Immunohistochemistry-Paraffin 1:200 - 1:500 |
| Zusammensetzung | PBS (pH 7.2), 40% Glycerol with 0.02% Sodium Azide |
| Gen-Alias | DKFZp781J1697, FLJ21629, KIAA0746FLJ41299, protein sel-1 homolog 3, sel-1 suppressor of lin-12-like 3 (C. elegans), Sel-1L3, Suppressor of lin-12-like protein 3 |
| Gensymbole | SEL1L3 |
| Wirtsspezies | Rabbit |
| Immunogen | This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:ERCAEVQEIVSVYASAAKHGGERQEACHLHNSYLDLQRRYGRPSMCRAFPWEKELKDKHPSLFQALLEMDLLTVPRNQNESVSEIGGKIFEKAVKRLSSIDGL |
| Show More |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?