Beschreibung
Semaphorin 4D/CD100 Polyclonal antibody specifically detects Semaphorin 4D/CD100 in Human samples. It is validated for Western Blot
Spezifikation
Spezifikation
| Antigen | Semaphorin 4D/CD100 |
| Anwendungen | Western Blot |
| Klassifikation | Polyclonal |
| Konjugat | Biotin |
| Zusammensetzung | PBS |
| Gen-Alias | A8, CD100 antigen, CD100M-sema-G, coll-4, FLJ33485, FLJ34282, FLJ39737, FLJ46484, immunoglobulin domain (Ig), transmembrane domain (TM) and shortcytoplasmic domain, 4D, MGC169138, MGC169141, sema domain, immunoglobulin domain (Ig), transmembrane domain (TM) and shortcytoplasmic domain, (semaphorin) 4D, semaphorin-4D |
| Wirtsspezies | Rabbit |
| Immunogen | Synthetic peptide corresponding to residues 777-788 [KPKSDFCDREQS] of the human CD100 protein. |
| Reinigungsverfahren | Immunogen affinity purified |
| Menge | 0.1 mL |
| Mehr anzeigen |