missing translation for 'onlineSavingsMsg'
Learn More

SEPX1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marke:  Proteintech 15333-1-AP-150UL

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 16884464

  • 329.85 € / 150 Mikroliter

Voraussichtliches Versanddatum: 25-06-2021

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

This gene encodes a selenoprotein, which contains a selenocysteine (Sec) residue at its active site. The selenocysteine is encoded by the UGA codon that normally signals translation termination. The 3′ UTR of selenoprotein genes have a common stem-loop structure, the sec insertion sequence (SECIS), that is necessary for the recognition of UGA as a Sec codon rather than as a stop signal. This protein belongs to the methionine sulfoxide reductase B (MsrB) family, and it is expressed in a variety of adult and fetal tissues.
Spezifikation

Spezifikation

SEPX1
0.33 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q52KJ8, Q9JLC3, Q9NZV6
HSPC270, MSRB1, Selenoprotein X, selenoprotein X, 1, SELR, SELX, SEPX1
Rabbit
IgG
150 μL
-20°C
Primary
27361, 51734, 685059
Immunohistochemistry (Paraffin)
Unconjugated
MSRB1
Liquid
Msrb1
SEPX1 Fusion Protein Ag7391
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Zertifikate