Learn More
Invitrogen™ SEZ6 Biosimilar Recombinant Monoclonal Antibody
GREENER_CHOICE
Artikelnummer. 30282407
Change view
Click to view available options
Menge:
1 mg
100 μg
Packungsgröße:
1 Milligramm
100 Mikrogramm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
30282407 100 μg 100 Mikrogramm
30284550 1 mg 1 Milligramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30282407 Lieferant Invitrogen™ Lieferanten-Nr. MA560101
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Recombinant Monoclonal Antibody
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °. This antibody can also be cross referenced by the term Abbv-011.
Sez6 may play a role in cell-cell recognition and in neuronal membrane signaling. Seems to be important for the achievement of the necessary balance between dendrite elongation and branching during the elaboration of a complex dendritic arbor. Involved in the development of appropriate excitatory synaptic connectivity.
TRUSTED_SUSTAINABILITY
Spezifikation
Antigen SEZ6 (Biosimilar)
Anwendungen ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance
Klassifikation Recombinant Monoclonal
Konzentration 1 mg/mL
Konjugat Unconjugated
Zusammensetzung 25mM histidine with 0.01% Tween 80, 8% sucrose and no preservative; pH 6.2
Gen Sez6
Gen-Zugriffsnummer Q53EL9
Gen-Alias brain-specific receptor-like protein C; BSRPC; BSRP-C; D11Bhm177e; hSEZ-6; Kiaa4158; Seizure protein 6; seizure protein 6 homolog; seizure related 6 homolog; seizure related 6 homolog (mouse); seizure related gene 6; SEZ6; SEZ-6
Gensymbole Sez6
Wirtsspezies Human
Reinigungsverfahren Protein A
Menge 100 μg
Regulatorischer Status RUO
Primär oder sekundär Primary
Gen-ID (Entrez) 124925
Zielspezies Human
Inhalt und Lagerung -20°C, Avoid Freeze/Thaw Cycles
Produkttyp Antibody
Form Lyophilized
Isotype IgG1 κ