missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SFRP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 26460-1-AP-20UL
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
This gene encodes a member of the SFRP family that contains a cysteine-rich domain homologous to the putative Wnt-binding site of Frizzled proteins. Members of this family act as soluble modulators of Wnt signaling; epigenetic silencing of SFRP genes leads to deregulated activation of the Wnt-pathway which is associated with cancer. This gene may also be involved in determining the polarity of photoreceptor cells in the retina.Spezifikation
SFRP1 | |
Polyclonal | |
Unconjugated | |
SFRP1 | |
FRP, FRP 1, FRP1, FrzA, SARP 2, SARP2, sFRP 1, SFRP1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
20377, 6422, 84402 | |
-20°C | |
Liquid |
Western Blot, Immunocytochemistry, Immunofluorescence | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q8C4U3, Q8N474, Q9R168 | |
SFRP1 | |
SFRP1 Fusion Protein Ag24844 | |
20 μL | |
Primary | |
Human, Rat, Mouse | |
Antibody | |
IgG |