missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ SID 26681509
Cathepsin L inhibitor
N.V.
Spezifikation
Reinheit | >97% |
---|---|
Produkttyp | SID 26681509 |
Molekulargewicht | 539.65 |
Hemmstoffe | SID 26681509 |
Beschreibung
Potent and reversible human cathepsin L inhibitor (IC50 = 56 nM). Displays no inhibitory activity at cathepsin G. Antimalarial; inhibits leishmaniasis.Spezifikation
>97% | |
SID 26681509 | |
539.65 | |
SID 26681509 |
For Research Use Only