Description
- PTFE septa is chemically resistant and allows puncturing
- Silicone lining allows repeated puncturing by preventing coring damage
- Slitted septa require less pressure to penetrate, preventing needle damage
- Packaged as 100 pieces (septa only) in a polyethylene bag
- Linerless black polypropylene screw caps with a center hole for type N8 vials are sold separately (mfr. no. 70249)
Specifications
Specifications
| Description | Hardness: 45° Shore A |
| Diameter (Metric) Cap | 8 mm |
| Color | Red, White |
| Diameter (Metric) Outer | 8 mm |
| Material | PTFE, Silicone |
| Pre-slit | No |
| For Use With (Application) | Chromatography (GC, HPLC) |
| Style | Round |
| Hardness | 45° Shore A |
| Quantity | 100 Pk. |
| Show More |