missing translation for 'onlineSavingsMsg'
Learn More

Silber(I)-fluorid, 98 %, Thermo Scientific Chemicals

Catalog Number p-4407842
Click to view available options
Menge:
5 g
25 g
7775-41-9
AgF
126.867
MFCD00003410
REYHXKZHIMGNSE-UHFFFAOYSA-M
silver fluoride, silver 1+ fluoride, silver 1+ ion fluoride, argentous fluoride, ag-f, silverfluoride, silver salt, fluoride, silver fluoride,, silver i fluoride 5g
165912
CHEBI:30340
Silber; Fluorid
[F-].[Ag+]
7775-41-9
AgF
126.867
MFCD00003410
REYHXKZHIMGNSE-UHFFFAOYSA-M
silver fluoride, silver 1+ fluoride, silver 1+ ion fluoride, argentous fluoride, ag-f, silverfluoride, silver salt, fluoride, silver fluoride,, silver i fluoride 5g
165912
CHEBI:30340
Silber; Fluorid
[F-].[Ag+]

Leicht, feuchtigkeitsempfindlich und hygroskopisch

Silver(I) fluoride is used as a fluorination and desilylation reagent in organic synthesis. In medical filed, it is used as topical caries in aqueous form in dentistry. It is employed in organofluorine chemistry for addition of fluoride across multiple bonds, especially to prepare perfluoroalkylsilver(I) by reacting with alkenes. Further, it is involved in the preparation of tetralkylammonium fluorides by the reaction with tetralkylammonium bromide. In addition to this, it is used as a desulfuration-fluorination reagent on thiourea derived substrates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Siedepunkt ca 1150°C
Chemischer Name oder Material Silver(I) fluoride
Schmelzpunkt 435°C
Farbe Weiß
Physikalische Form Kristallin
Menge 5 g
Prozentgehaltsbereich 98%
Löslichkeitsinformationen Soluble in HF,ammonia,CH3CNSoluble in water,hydrogen fluoride and methanol.
Formelmasse 126.87
Empfindlichkeit Hygroscopic; Light and moisture sensitive
Aussehen Orange-brown crystals
Dichte 5.852 g/cm3 at 20°C
Mehr anzeigen Weniger anzeigen

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'