missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Silbernitrat, 1.0 N standardisierte Lösung, Thermo Scientific Chemicals
Click to view available options
Menge:
500 mL
1 L
Beschreibung
Silver nitrate, 1.0N Standardized Solution is used for volumetric analysis in laboratories. It is widely used as an analytical reagent owing to its immediate reaction with halide ions forming insoluble precipitate of silver(I) halides.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Spezifikation
Chemischer Name oder Material | Silver nitrate |
Farbe | Farblos |
Physikalische Form | Flüssig |
Menge | 500 mL |
Löslichkeitsinformationen | Miscible with water,alcohol and aqueous ammonia. |
Formelmasse | 169.87 |
Empfindlichkeit | Light sensitive |
Aussehen | Colorless liquid |
Dampfdruck | 23 hPa (17mm Hg) at 20°C |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'