missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Simport™ Scientific OneHand™ Mikroröhrchengestell
N.V.
Spezifikation
Autoklavierbar | Not Autoclavable |
---|---|
Abmessungen (L x B x H) | 11.5 x 4.5 x 1.5 in. (29.3 x 11.45 x 3.9cm) |
Material | Acetal Polymer |
Positionen | 50 |
Menge | 10/Cs. |
Spezifikation
Not Autoclavable | |
Acetal Polymer | |
10/Cs. |
11.5 x 4.5 x 1.5 in. (29.3 x 11.45 x 3.9cm) | |
50 |