Learn More
IGF1 Recombinant Human Protein, His.NusA Tag, Invitrogen™ Sino Biological™
This recombinant protein was expressed from a DNA sequence encoding the mature form of human IGF1 isoform 1 (P05019-1) (Gly 49-Ala 118) fused to the polyhistidine-NusA tag at the N-terminus
Marke: Invitrogen 10598H24E5
Weitere Details : Netto-Gewicht : 0.00001kg
Beschreibung
Insulin-like growth factor 1 (IGF1), recombinant human protein is supplied as a lyophilized powder. In general, recombinant proteins can be used as protein standard and in cell biology research applications.- Activity: Measured by its binding ability in a functional ELISA; immobilized human IGF1 at 10μg/mL (100μL/well) can bind biotinylated human IGFBP3-His, the EC50 of biotinylated human IGFBP3-His is 24ng/mL
- Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
- Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible
This protein is manufactured by Sino Biological Inc.
Spezifikation
Protein Expression and Isolation, Enzyme and Protein Activity Assay, Protein Assay Control, Reference Standards and Accessories | |
5μg | |
Full Length/Recombinant | |
Insulin-like growth factor 1 (somatomedin C) | |
His Tag, NusA Tag | |
Growth Factors & Receptors | |
IGFs (Insulin-Like Growth Factors) |
64.8kDa | |
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed. | |
IGF-I, IGF1A, IGFI | |
IGF1 | |
E. coli | |
Lyophilized | |
≥90% by SDS-PAGE |
For Research Use Only. Not for use in diagnostic procedures.