missing translation for 'onlineSavingsMsg'
Learn More

IL12A and IL12B Recombinant Human Protein, His Tag, Invitrogen™ Sino Biological™

This recombinant heterodimer protein was expressed from a DNA sequence encoding the p35 subunit of human IL-12, termed as IL12A (NP_000873.2) (Met 1-Ser 219) fused to a polyhistidine tag at the C-terminus, constructed the plasmid 1, and from a DNA sequence

Marke:  Invitrogen CT011H08H5

Weitere Details : Netto-Gewicht : 0.01000kg

Artikelnummer. 15431114

Dieser Artikel ist im Moment nicht verfügbar oder wird nicht mehr hergestellt.
Schauen Sie bitte auf unsere Produktseite nach möglichen Alternativen


Beinhaltet: One vial of lyophilized protein

Beschreibung

Beschreibung

Interleukin 12 (Interleukin 12A and Interleukin 12B Heterodimer) (IL12A and IL12B), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its ability to bind biotinylated recombinant human IL12RB1 in a functional ELISA
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Spezifikation

Spezifikation

Assay Standard, Enzyme and Protein Activity Assay, Protein Assay Control, Reference Standards and Accessories
5μg
<1 EU/μg
Interleukin 12 (interleukin 12A and interleukin 12B heterodimer)
His Tag
Cytokines & Receptors
Interleukins
60.2kDa
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Interleukin-12
IL12A and IL12B
Mammalian
Lyophilized
≥90% by SDS-PAGE
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote

For Research Use Only. Not for use in diagnostic procedures.