missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S100A9 Recombinant Human Protein, Invitrogen™ Sino Biological™
This recombinant protein was expressed from a DNA sequence encoding the human S100A9 (NP_002956.1) (Met 1-Pro 114)
299.00 € - 945.00 €
Spezifikation
Zur Verwendung mit (Anwendung) | Protein Expression and Isolation, Enzyme and Protein Activity Assay, Protein Assay Control, Reference Standards and Accessories |
---|---|
Molekulargewicht | 26kDa |
Lagerungsbedingungen | Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed. |
Gen-Alias | 60B8AG, CAGB, CFAG, CGLB, L1AG, LIAG, MAC387, MIF, MRP14, NIF, P14 |
Gebräuchliche Bezeichnung | S100 calcium binding protein A9 |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15248305
|
Invitrogen
11145HNAE50 |
50μg |
Dieser Artikel ist im Moment nicht verfügbar oder wird nicht mehr hergestellt. Schauen Sie bitte auf unsere Produktseite nach möglichen Alternativen |
||||||
15228305
|
Invitrogen
11145HNAE250 |
5 x 50μg |
Dieser Artikel ist im Moment nicht verfügbar oder wird nicht mehr hergestellt. Schauen Sie bitte auf unsere Produktseite nach möglichen Alternativen |
||||||
Beschreibung
S100 Calcium Binding Protein A9 (S100A9), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, and performing cell based assays. It can also be used as an immunogen, as a protein standard, or in other research applications.- Activity: Measured by its ability to inhibit the proliferation of MCF7 human breast adenocarcinoma cells; the ED50 for this effect is typically 3-15μg/mL
- Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
- Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible
This protein is manufactured by Sino Biological Inc.
Spezifikation
Protein Expression and Isolation, Enzyme and Protein Activity Assay, Protein Assay Control, Reference Standards and Accessories | |
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed. | |
S100 calcium binding protein A9 | |
Untagged | |
Other Proteins | |
Other Proteins |
26kDa | |
60B8AG, CAGB, CFAG, CGLB, L1AG, LIAG, MAC387, MIF, MRP14, NIF, P14 | |
S100A9 | |
E. coli | |
Lyophilized | |
≥90% by SDS-PAGE |
For Research Use Only. Not for use in diagnostic procedures.