missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ SULT1A1 Recombinant Human Protein, N-His Tag

This recombinant protein was expressed from a DNA sequence encoding the human SULT1A1 (P50225-1) (Glu 2-Leu 295) fused to a polyhistidine tag at the N-terminus

Marke:  Thermo Scientific™ 11658H07E5

Weitere Details : Netto-Gewicht : 0.01000kg

Artikelnummer. 15288795

Dieser Artikel ist im Moment nicht verfügbar oder wird nicht mehr hergestellt.
Schauen Sie bitte auf unsere Produktseite nach möglichen Alternativen


Beschreibung

Beschreibung

Sulfotransferase Family, Cytolsolic, 1A, Phenol-Preferring, Member 1 (SULT1A1), recombinant human protein is supplied as a frozen protein. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: 50mM Tris, 150mM NaCl, 10% Glycerol, pH 8.0
  • Reconstitution: No reconstitution needed

This protein is manufactured by Sino Biological Inc.

Spezifikation

Spezifikation

Protein Expression and Isolation, Enzyme and Protein Activity Assay, Protein Assay Control, Reference Standards and Accessories
35kDa
5μg
Full Length/Recombinant
Sulfotransferase Family, Cytolsolic, 1A, Phenol-Preferring, Member 1
His Tag
Other Proteins
≥90% by SDS-PAGE
6817
PAGE
Store at -80°C. Thaw protein on ice before use and avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
OK/SW-cl.88, HAST1/HAST2, mgC131921, mgC5163, P-PST, PST, ST1A1, ST1A3, STP, STP1, TSPST1
SULT1A1
E. coli
Frozen
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote

For Research Use Only. Not for use in diagnostic procedures.