missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SIRT5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 15122-1-AP-150UL
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
SIRT5 is a human member of a family of proteins called Sirtuins (Sir2-like proteins) and are present in prokaryotes and eukaryotes. All Sir2-like proteins have a sirtuin core domain, which contains a series of sequence motifs conserved in organisms ranging from bacteria to humans. Bacterial, yeast and mammalian sirtuins are able to metabolize NAD and possibly at as mono-ADP-ribosyltransferases. The enzymatic function of sirtuins is not yet completely understood but recent reports of histone-activated Sir2-mediated NAD metabolism and NAD-activated Sir2-mediated histone deacetylation suggest a possible coupled reciprocal activation mechanism involving interactions of Sir2 with NAD and the N epsilon-acetyl-lysine groups of acetylated histones.Spezifikation
SIRT5 | |
Polyclonal | |
Unconjugated | |
SIRT5 | |
SIR2 like protein 5, SIR2L5, SIRT5 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
23408, 306840, 68346 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q68FX9, Q8K2C6, Q9NXA8 | |
Sirt5 | |
SIRT5 Fusion Protein Ag7195 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |