Learn More
Tocris Bioscience™ SIS3
Selective Smad3 inhibitor; inhibits TGF-βR1 signaling
N.V.
Spezifikation
Produkttyp | SIS3 |
---|---|
Beschreibung | SIS3 |
Molekulargewicht | 489.99 |
Beschreibung
Selective Smad3 inhibitor; attenuates TGF-β1-dependent Smad3 phosphorylation and DNA binding. Has no effect on Smad2, p38 MAPK, ERK or PI 3-kinase signaling. Inhibits TGF-β1-induced myofibroblast differentiation of dermal fibroblasts. Also inhibits TGF-β2-induced endothelial cell differentiation in iPSCs.Spezifikation
SIS3 | |
SIS3 | |
489.99 |
For Research Use Only