missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ SKA 31
KCa3.1 (IK) and KCa2 (SK) channel activator
N.V.
Spezifikation
Reinheit | >99% |
---|---|
Produkttyp | SKA 31 |
Molekulargewicht | 200.26 |
Hemmstoffe | SKA 31 |
Beschreibung
Activator of KCa3.1 and KCa2 channels (EC50 values are 260, 2900, 2900 nM for KCa3.1, KCa2.1 and KCa2.2 respectively). Potentiates acetylcholine-induced EDHF-type dilations of mouse carotid arteries and lowers blood pressure in normotensive and hypertensive mice.Spezifikation
>99% | |
SKA 31 | |
200.26 | |
SKA 31 |
For Research Use Only