Learn More
B Braun™ Michel Tissue Forceps
Product Code. 10535055
Change view
Click to view available options
Length (Metric):
125 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric)
10535055 125 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10535055 Supplier B Braun™ Supplier No. BN750R
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
This item is not returnable. View return policy
B Braun™ Michel Tissue Forceps are commonly used for applying Michel's clips or skin sutures.
  • Clip applying and removing forceps
  • Stainless steel construction
Specifications
Product Type Michel Clip Applying/Removing
Length (Metric) 125 mm
Material Stainless Steel