missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Slc17a8 Mouse anti-Human, Rat, HRP, Clone:S34-34, Abnova™
Specifications
Specifications
| Antigen | Slc17a8 |
| Anwendungen | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot |
| Klassifikation | Monoclonal |
| Klon | S34-34 |
| Konjugat | HRP |
| Beschreibung | solute carrier family 17 (sodium-dependent inorganic phosphate cotransporter), member 8 |
| Verdünnung | Immunocytochemistry (1:100) Immunofluorescence (1:100) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) Western Blot (1:1000) The optimal working dilution should be determined by the end user. |
| Zusammensetzung | In PBS, pH 7.4 (50% glycerol, 0.09% sodium azide) |
| Gen | Slc17a8 |
| Gen-Alias | Vglut3 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?