missing translation for 'onlineSavingsMsg'
Learn More

SLC22A1 Antibody (2C5), Novus Biologicals™

Artikelnummer. 18506034 Alle ansehen Bio Techne Produkte
Change view
Click to view available options
Menge:
0.025 mL
Packungsgröße:
25 Mikroliter
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
18506034 0.025 mL 25 Mikroliter
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 18506034 Lieferant Novus Biologicals Lieferanten-Nr. NBP1516840.025ml

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Mouse Monoclonal Antibody has been used in 6 publications

SLC22A1 Monoclonal specifically detects SLC22A1 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Electron Microscopy.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC22A1
Anwendungen Western Blot
Klassifikation Monoclonal
Klon 2C5
Konjugat Unconjugated
Verdünnung Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Electron Microscopy
Gen-Zugriffsnummer O15245
Gen-Alias hOCT1, oct1_cds, OCT1solute carrier family 22 member 1, Organic cation transporter 1, solute carrier family 22 (organic cation transporter), member 1
Gensymbole SLC22A1
Wirtsspezies Mouse
Immunogen Purified recombinant fragment of human SLC22A1 expressed in E. Coli.
Molekulargewicht des Antigens 61.2 kDa
Reinigungsverfahren Unpurified
Menge 0.025 mL
Regulatorischer Status RUO
Primär oder sekundär Primary
Gen-ID (Entrez) 6580
Zielspezies Human
Inhalt und Lagerung Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.
Form Ascites
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.