Learn More
SLC25A3 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke:  Proteintech 10420-1-AP-20UL
Weitere Details : Netto-Gewicht : 0.00010kg
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 16886793
  • 112.71 € / 20 Mikroliter
Verfügbar ab
17-06-2021
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Beschreibung
Beschreibung
The protein encoded by this gene catalyzes the transport of phosphate into the mitochondrial matrix, either by proton cotransport or in exchange for hydroxyl ions. The protein contains three related segments arranged in tandem which are related to those found in other characterized members of the mitochondrial carrier family. Both the N-terminal and C-terminal regions of this protein protrude toward the cytosol.
Spezifikation
Spezifikation
SLC25A3
0.23 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q00325
OK/SW cl.48, PHC, Phosphate transport protein, PiC, PTP, SLC25A3
Rabbit
IgG
20 μL
-20°C
Primary
5250
Western Blot, Immunohistochemistry (Paraffin)
Unconjugated
SLC25A3
Liquid
SLC25A3
SLC25A3 Fusion Protein Ag0667
Antigen Affinity Chromatography