missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC25A34 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 17557-1-AP-150UL
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
SLC25A34 belongs to the SLC25 family of mitochondrial carrier proteins.Spezifikation
SLC25A34 | |
Polyclonal | |
Unconjugated | |
SLC25A34 | |
RP11 169K16.2, SLC25A34 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
284723, 298606, 384071 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
A2ADF7, Q5XIF9, Q6PIV7 | |
SLC25A34 | |
SLC25A34 Fusion Protein Ag11719 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |