Learn More
SLC3A1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 16343-1-AP-20UL
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
This gene encodes a type II membrane glycoprotein which is one of the components of the renal amino acid transporter which transports neutral and basic amino acids in the renal tubule and intestinal tract. Mutations and deletions in this gene are associated with cystinuria. Alternatively spliced transcript variants have been described, but their biological validity has not been determined.Spezifikation
SLC3A1 | |
Polyclonal | |
Unconjugated | |
SLC3A1 | |
ATR1, CSNU1, D2H, NBAT, RBAT, SLC3A1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
20532, 29484, 6519 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunoprecipitation | |
0.21 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q07837, Q64319, Q91WV7 | |
SLC3A1 | |
SLC3A1 Fusion Protein Ag9525 | |
20 μL | |
Primary | |
Mouse, Human, Rat | |
Antibody | |
IgG |