Learn More
Ibidi μ-Slide 8 Well high μ-Pattern RGD, sqr20, pit110, hex
μ-Slide 8 Well high μ-Pattern RGD, sqr20, pit110, hex is an 8 well chambered coverslip with micropatterned surface.
Marke: Ibidi 83801
Weitere Details : Netto-Gewicht : 0.57000kg
Beschreibung
- A chambered coverslip with 8 wells and micropatterned surface
- bottom material: #1.5 Polymer Coverslip with passivated surface (biologically inert) and RGD pattern
- sterilized
Spezifikation
Microscope Slide | |
8 | |
10 |