Learn More
Ibidi μ-Slide I Luer 3D ibiTreat
A μ-Slide has one channel and three wells.
Marke: Ibidi 87176
Weitere Details : Netto-Gewicht : 0.14300kg
Beschreibung
- A μ-Slide with one channel and three wells for culturing cells on a 3D gel matrix with defined flow
- easy to connect with tubing and pump systems via female Luers
- bottom material: #1.5 Polymer Coverslip
- tissue culture treated
- sterilized
Spezifikation
Microscope Slide | |
15 |