Learn More
Ibidi μ-Slide I Luer 3D Uncoated
Product Code. 17236263
Encompass_Preferred
Change view
Click to view available options
μ-Slide
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
17236263 μ-Slide
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17236263 Supplier Ibidi Supplier No. 87171
 Restricted Item
Verfügbar ab: 02-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
A μ-Slide has one channel and three wells.
  • A μ-Slide with one channel and three wells for culturing cells on a 3D gel matrix with defined flow
  • easy to connect with tubing and pump systems via female Luers
  • bottom material: #1.5 Polymer Coverslip
  • hydrophobic
  • sterilized
Specifications
Type Microscope Slide
Quantity 15