Learn More
Ibidi μ-Slide III 3D Perfusion Uncoated
Product Code. 15561255
Encompass_Preferred
Change view
Click to view available options
μ-Slide
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
15561255 μ-Slide
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15561255 Supplier Ibidi Supplier No. 80371
 Restricted Item
Verfügbar ab: 07-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
μ-Slide III 3D Perfusion Uncoated is a three channel μ-Slide used for the cultivation and perfusion of 3D cell structure.
  • A three channel slide used for the cultivation and perfusion of cells or spheroids in 3D matrices
  • easy to connect with tubing and pump systems via female Luers
  • bottom material: #1.5 Polymer Coverslip
  • hydrophobic
  • sterilized
Specifications
Type Microscope Slide
Quantity 15