Learn More
Ibidi μ-Slide VI 0.4 μ-Pattern RGD, sqr20, pit110, hex Trial Pack
μ-Slide VI 0.4 μ-Pattern RGD, sqr20, pit110, hex Trial Pack is a 6 channel slide with micropatterned surface.
Marke: Ibidi 83601-S
Weitere Details : Netto-Gewicht : 0.18600kg
Beschreibung
- A 6 channel slide with micropatterned surface
- bottom material: #1.5 Polymer Coverslip with passivated surface (biologically inert) and RGD pattern
- sterilized
Specifications
Microscope Slide | |
2 |