Learn More
Thermo Scientific™ Smart NIR Integrating Sphere Accessories
Product Code. 18875473 Shop All Thermo Scientific Products
Change view
Click to view available options
Type:
Sample Spinner
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Unit Size
18875473 Sample Spinner Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18875473 Supplier Thermo Scientific™ Supplier No. 840095700
  • ONLINE EXCLUSIVE PRICE
    4305.00 €
    4050.00 € / 1 Stück
    Sparen 255.00 €
    5% Rabatt
Verfügbar ab: 21-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
This item is not returnable. View return policy
Accessories for the Thermo Scientific™ Smart NIR Integrating Sphere accessory.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Smart NIR Integrating Sphere
Type Sample Spinner
For Use With (Application) Inhomogenous samples
Includes 5cm open powder sampling cup
Optics Quartz window