missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SMPX Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 15791-1-AP-150UL
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
SMPX plays a role in the regulatory network through which muscle cells coordinate their structural and functional states during growth, adaptation, and repair.Spezifikation
SMPX | |
Polyclonal | |
Unconjugated | |
SMPX | |
small muscle protein, X linked, Small muscular protein, SMPX, SRMX | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
23676, 66106, 84416 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.16 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q925F0, Q9DC77, Q9UHP9 | |
Smpx | |
SMPX Fusion Protein Ag8497 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |