missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SNURPORTIN1 Antibody (SNUPN/7363R), Allophycocyanin, Novus Biologicals™
Alle ansehen Bio Techne ProdukteBeschreibung
SNURPORTIN1 Monoclonal antibody specifically detects SNURPORTIN1 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Spezifikation
Spezifikation
| Antigen | SNURPORTIN1 |
| Anwendungen | Immunohistochemistry (Paraffin) |
| Klassifikation | Monoclonal |
| Klon | SNUPN/7363R |
| Konjugat | APC |
| Zusammensetzung | PBS |
| Gen-Alias | KPNBL, RNA U transporter 1, RNA, U transporter 1, RNUT1, snurportin 1, Snurportin1, SNURPORTIN-1, SPN1 |
| Wirtsspezies | Rabbit |
| Immunogen | Recombinant fragment (around aa 900-1100) of human SNURPORTIN1 protein (exact sequence is proprietary) |
| Reinigungsverfahren | Protein A or G purified |
| Mehr anzeigen |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?