missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SNW1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 25926-1-AP-20UL
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
CSTF2 is one of three (including CSTF1 and CSTF3) cleavage stimulation factors which combine to form CSTF which is involved in the polyadenylation and 3′end cleavage of pre-mRNAs. CSTF2 contains a ribonucleoprotein-type RNA binding domain. CSTF2 is upregulated during activation of B cells which results in the switch of IgM heavy chain mRNA from membrane bound form to the secreted form.Spezifikation
SNW1 | |
Polyclonal | |
Unconjugated | |
SNW1 | |
Bx42, NCOA 62, Nuclear protein SkiP, Prp45, PRPF45, Ski interacting protein, SKIIP, SKIP, SNW domain containing 1, SNW1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
22938 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Western Blot | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q13573 | |
SNW1 | |
SNW1 Fusion Protein Ag23106 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |