missing translation for 'onlineSavingsMsg'
Learn More

Respiratory Syncytial Virus Subgroup B Antibody (4-14), R&D Systems™

Product Code. 18606536 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
Unit Size:
0.03mL
0.1mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18606536 0.025 mL 0.03mL
18632829 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18606536 Supplier Novus Biologicals Supplier No. NBP2503110.025ml
 Restricted Item
Verfügbar ab: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Novus product, part of R&D Systems. Mouse Monoclonal Antibody

Respiratory Syncytial Virus Subgroup B Monoclonal specifically detects Respiratory Syncytial Virus Subgroup B in Virus samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence, Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Respiratory Syncytial Virus Subgroup B
Applications ELISA, Immunocytochemistry, Immunofluorescence, Western Blot
Classification Monoclonal
Clone 2024-04-14 00:00:00
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500
Formulation PBS
Host Species Mouse
Immunogen The immunogen was gradient-purified RSN-2 virus (subgroup B) that was then treated with 0.1% SDS at 100C for 2 min. The procedure used to produce this antibody is described in Gimenez et al. (1984).
Purification Method Thiophilic Chromatography
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Virology Bacteria and Parasites
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Product Type Antibody
Form Purified
Isotype IgM
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.