missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Hydroxide, 5.0N Solution, Water for Injection Quality Water, Biotech Reagent, J.T. Baker™
Click to view available options
Menge:
19 L
Verpackung:
Poly Bottle
Spezifikation
Chemischer Name oder Material | Sodium Hydroxide, 5.0N Solution |
Namenshinweis | Water for Injection Quality Water |
CAS | 1310-73-2 |
Summenformel | HNaO |
Synonym | sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic |
InChI-Schlüssel | HEMHJVSKTPXQMS-UHFFFAOYSA-M |
Menge | 19 L |
SMILES | [OH-].[Na+] |
IUPAC-Name | sodium;hydroxide |
Molekulargewicht (g/mol) | 39.997 |
Mehr anzeigen |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'