missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium hydroxide solution, ≈0.1M, Honeywell Fluka™
Click to view available options
Menge:
100 mL
500 mL
Spezifikation
Chemischer Name oder Material | Sodium hydroxide solution |
Strukturformel | NaOH |
Konzentration | ≈0.1M |
UN-Nummer | NONH for all modes of transport |
Menge | 100 mL |
Explosionsgrenze | for HPCE |
Formelmasse | 40g/mol |
Dampfdruck | 3 mmHg (37°C) |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'