Learn More
Sodium Nitrite, TRC
Product Code. 30422602
Change view
Click to view available options
Quantity:
250 g
Unit Size:
250g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30422602 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30422602 Supplier Toronto Research Chemicals Supplier No. TRCS648000250G
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Sodium nitrite
CAS 7632-00-0
Quantity 250 g
Molecular Formula N O2 . Na
Synonym Sodium nitrite, Nitrous Acid Sodium Salt, Anti-Rust, E 250, Erinitrit, Filmerine, M 138C, Nitrous Acid Soda, SNS 40, Sodium Nitrite, Sodium Nitrite (NaNO2), Synfat 1004
InChI Formula InChI=1S/HNO2.Na/c2-1-3;/h(H,2,3);/q;+1/p-1
SMILES [Na+].[O-]N=O
IUPAC Name sodium;nitrite
Molecular Weight (g/mol) 69
Formula Weight 68.9827