missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Somatostatin Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 17512-1-AP-150UL
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
This antibody can recognize the full length and propeptide of Somatostatin.
The hormone somatostatin has active 14 aa and 28 aa forms that are produced by alternate cleavage of the single preproprotein encoded by this gene. Somatostatin is expressed throughout the body and inhibits the release of numerous secondary hormones by binding to high-affinity G-protein-coupled somatostatin receptors. This hormone is an important regulator of the endocrine system through its interactions with pituitary growth hormone, thyroid stimulating hormone, and most hormones of the gastrointestinal tract. Somatostatin also affects rates of neurotransmission in the central nervous system and proliferation of both normal and tumorigenic cells.Spezifikation
Somatostatin | |
Polyclonal | |
Unconjugated | |
SST | |
SMST, somatostatin, SST | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
20604, 6750 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.31 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P60041, P61278 | |
SST | |
Peptide | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |