missing translation for 'onlineSavingsMsg'
Learn More

SPATA4 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Marke:  Thermo Scientific PA553959

Weitere Details : Netto-Gewicht : 0.02000kg

Artikelnummer. 15771891

  • 477.00 € / 100 Mikroliter
Verfügbar ab: 23-04-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

Immunogen sequence: LILLHMLQRK LGRKLNPEWF DVKPTVGEVT LNHLPAQASG RRYNLKVKRG RVVPVLPNIG SGGSSHREIH VKQAGQHSYY SAMKPIRN Highest antigen sequence identity to the following orthologs: Mouse - 47%, Rat - 55%.

SPATA4 Polyclonal antibody specifically detects SPATA4 in Human samples. It is validated for Immunohistochemistry.
Spezifikation

Spezifikation

SPATA4
Polyclonal
Unconjugated
SPATA4
1700001N01Rik; Spata4; SPEF1B; spermatogenesis associated 4; spermatogenesis related gene 2; spermatogenesis-associated 4; spermatogenesis-associated protein 4; Spermatogenesis-related gene 2 protein; Srg2; testis and spermatogenesis cell related protein 2; testis and spermatogenesis cell-related protein 2; Testis spermatocyte apoptosis-related gene 2 protein; TSARG2
Rabbit
Antigen affinity chromatography
RUO
132851
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry
0.3 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q8NEY3
SPATA4
Recombinant protein corresponding to Human SPATA4
100 μL
Primary
Human
Antibody
IgG
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote

Research Use Only