Learn More
Beckman Coulter SPRIselect DNA Size Selection Reagent
Artikelnummer. 31723109
Change view
Click to view available options
Fassungsvermögen:
5 mL
60 mL
450 mL
Packungsgröße:
450 Milliliter
5 Milliliter
60 Milliliter
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Fassungsvermögen unitSize
31723109 5 mL 5 Milliliter
31723148 60 mL 60 Milliliter
31723147 450 mL 450 Milliliter
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Refer to SDS
Artikelnummer. 31723109 Lieferant Beckman Coulter Lieferanten-Nr. B23317
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Refer to SDS
Bead-based reagent for DNA size selection and cleanup.
  • SPRI select Size Selection reagent kit is a SPRI-based chemistry that speeds and simplifies nucleic acid size selection for fragment library preparation for next generation sequencing (NGS).
  • SPRI select yields consistent size selections without bead calibration between reagent lots. Suitable for manual or automated workflows - the SPRI select method can be run on a variety of Biomek platforms that offer minimal hands-on time.
TRUSTED_SUSTAINABILITY
Spezifikation
Fassungsvermögen 5 mL
Isolation Technology Magnetic Bead
Produkttyp DNA Size Selection Reagent