Learn More
SRMS (SRM) Recombinant Human Protein
SRMS (SRM) Recombinant Human Protein
Marke:  Invitrogen PV4214
Weitere Details : Netto-Gewicht : 0.05250kg
Artikelnummer. 11543637
  • 632.00 € / 10 Mikrogramm
Verfügbar ab: 16-04-2024
Sie sich ein, um den Lagerbestand zu sehen
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Nur für Forschungszwecke. Die Anwendung muss den Produktanweisungen entsprechen.
Beschreibung
Beschreibung
SRMS (Srm) is a tyrosine protein kinase (SRC sub-family) that may be involved in the proliferation and differentiation of keratinocytes.
Spezifikation
Spezifikation
NP_543013
Pharma and Biopharma, Protein Assays and Analysis, Target and Lead Identification and Validation, Kinase Assay
82kDa
10μg
For research use only. Not for use in diagnostic procedures
SRMS (Srm)
SRMS (Srm) was pre-diluted in enzyme dilution buffer and assayed in 25mM HEPES (pH 7.5), 10mM MgCl2, 0.5mM EGTA, 0.5mM Na3VO4, 5mM β-glycerophosphate, 2.5mM DTT, 0.01% Triton™ X-100, 200μM ATP, 200μg/mL poly [Glu, Tyr] 4:1 substrate and trace [32P]-γ-ATP for 10 minutes at 30°C
SRMS (SRM) Recombinant Human Protein
Human
Baculovirus
Cytoplasmic tyrosine kinase
0.19mg/mL
114294, 6725
SRMS (SRM) Recombinant Human Protein
–80°C
1 Each
SRMS (SRM) Recombinant Human Protein
SRC
130nmoles of phosphate transferred to poly [Glu, Tyr] 4:1 substrate per minute per mg of total protein at 30°C. Activity determined at a final protein concentration of 10μg/mL
GST Tag
Kinase and inhibitors
90% by SDS-PAGE