missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SRPK1 Recombinant Human Protein
SRPK1 is a serine/threonine kinase.
Marke: Invitrogen™ PR7847A
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
SRPK1 plays a role in the regulation of both constitutive and alternative splicing by regulating intracellular localization of splicing factors.
Spezifikation
NP_003128 | |
50mM Tris-HCl pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% Glycerol | |
100μg | |
RUO - research use only | |
SRPK1, SFRSK1 | |
Human | |
His Tag | |
Frozen |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
78.4kDa | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Full Length | |
SRPK Kinase Family | |
Recombinant | |
Baculovirus |