missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SSX4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA550833
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
The antibody detects endogenous levels of total SSX4 protein.
SSX4 Polyclonal antibody specifically detects SSX4 in Human samples. It is validated for Immunohistochemistry (Paraffin).Spezifikation
SSX4 | |
Polyclonal | |
Unconjugated | |
SSX4 | |
cancer/testis antigen 5.4; CT5.4; MGC119056; MGC12411; Protein SSX4; SSX family member 4; SSX4; SSX4A; SSX4B; synovial sarcoma, X breakpoint 4 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
6759 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
1.1 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
O60224 | |
SSX4 | |
Synthetic peptide corresponding to residues near the C terminal of human synovial sarcoma, X breakpoint 4 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |