Learn More
Novus Biologicals™ ST8 alpha-2,8-Sialyltransferase 8A/ST8SIA1/Ganglioside GD3 Overexpression Lysate
Alle ansehen Bio Techne ProdukteBeschreibung
The ST8 alpha-2,8-Sialyltransferase 8A/ST8SIA1/Ganglioside GD3 Overexpression Lysate is derived from human. It has been validated for the following applications: Western Blot.
Spezifikation
Spezifikation
| Zelllinie | The lysate was created in HEK293T cells, using plasmid ID RC223851 and based on accession number NM_334. The protein contains a C-terminal DDK tag. |
| Zur Verwendung mit (Anwendung) | Western Blot |
| Menge | 0,1 mg |
| Lagerungspuffer | RIPA buffer |
| Reinheit | Protein |
| Testspezifität | Homo sapiens ST8 alpha-N-acetyl-neuraminide alpha-2,8-sialyltransferase 1 (ST8SIA1), mRNA. |
| Inhalt und Lagerung | -80°C. Avoid freeze-thaw cycles. |
| Protein State | Native |
Nur für Forschungszwecke