Learn More
Edelstahlpulver, -20 +35 Mesh, Typ 316-L, Thermo Scientific Chemicals
Beschreibung
Stainless Steel gauze, Type 316 is used to woven wire cloth which has extensive use in chemical processing when better corrosion resistance is required than the regular chromium-nickel types. It is also used in medical, pharmaceutical, laboratory, semiconductor, food processing, marine, waste water and water purification equipment.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Spezifikation
Spezifikation
| Chemischer Name oder Material | Stainless Steel powder, Type 316-L |
| Durchschnitt Mol. Gew. oder Mol.-Gew. Gewichtsbereich | Fe:Cr:Ni:Mo; 67.5:17:13:2.5 wt% |
| Menge | 500 g |
| Netzgröße | −20+35 Mesh |
| Form | Pulver |
| MDL-Nummer | MFCD00148499 |
| Löslichkeitsinformationen | Insoluble in water. |